Skip to Content
Merck
All Photos(1)

Key Documents

860592P

Avanti

24:0 SM

Avanti Research - A Croda Brand 860592P, powder

Synonym(s):

N-lignoceroyl-D-erythro-sphingosylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H95N2O6P
CAS Number:
Molecular Weight:
815.24
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860592P-10mg)
pkg of 1 × 5 mg (860592P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860592P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C47H95N2O6P/c1-6-8-10-12-14-16-18-20-21-22-23-24-25-26-27-29-31-33-35-37-39-41-47(51)48-45(44-55-56(52,53)54-43-42-49(3,4)5)46(50)40-38-36-34-32-30-28-19-17-15-13-11-9-7-2/h38,40,45-46,50H,6-37,39,41-44H2,1-5H3,(H-,48,51,52,53)/b40-38+/t45-,46+/m0/s1

InChI key

QEDPUVGSSDPBMD-XTAIVQBESA-N

General description

d-erythro (2S, 3R) sphingomyelin (C24:0-SM) is a synthetic sphingomyelin with acyl chain comprising 24 carbon atom. It is produced by the acylation of D-erythro-sphingosylphosphorylcholine. Sphingomyelin is a key constituent of mammalian cell plasma membranes. It is mainly found at high concentrations in the myelin sheaths of nerve cells, in brain, eye lens and red blood cells. SM have asymmetric molecular structure and high hydrogen-bonding capacity.

Application

24:0 sphingomyelin (SM) is suitable to study its thermotropic and mechanical properties by using differential scanning calorimetry. It is also suitable for use as a lipid standard in ultrafast liquid chromatography−tandem mass spectrometry.

Biochem/physiol Actions

Sphingomyelin (SM) are involved in the formation of functional domains in the membrane bilayer. It also acts as a precursor for sphingolipid signaling molecules including, ceramides and other sphingolipid metabolites.

Packaging

5 mL Amber Glass Screw Cap Vial (860592P-10mg)
5 mL Amber Glass Screw Cap Vial (860592P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanical properties of milk sphingomyelin bilayer membranes in the gel phase: Effects of naturally complex heterogeneity, saturation and acyl chain length investigated on liposomes using AFM
Et-Thakafy O, et al.
Chemistry and Physics of Lipids, 210, 47-59 (2018)
Phase transitions of highly asymmetric chain-length N-lignocerylsphingomyelin (C24: 0-SM) bilayer
Ohtaka H, et al.
Journal of Thermal Analysis and Calorimetry, 113(3), 1593-1602 (2013)
Very-long-chain fatty acid sphingomyelin in nuclear lipid microdomains of hepatocytes and hepatoma cells: can the exchange from C24: 0 to C16: 0 affect signal proteins and vitamin D receptor?
Lazzarini A, et al.
Molecular Biology of the Cell, 26(13), 2418-2425 (2015)
Elisabetta Albi et al.
International journal of molecular sciences, 15(7), 11555-11565 (2014-07-01)
Proliferating thyroid cells are more sensitive to UV-C radiations than quiescent cells. The effect is mediated by nuclear phosphatidylcholine and sphingomyelin metabolism. It was demonstrated that proton beams arrest cell growth and stimulate apoptosis but until now there have been
Ebru Afşar et al.
Redox biology, 30, 101430-101430 (2020-01-25)
We aimed to determine sphingolipid levels and examine apoptotic pathways in human retinal pigment epithelial cells (ARPE-19) undergoing endoplasmic reticulum (ER) stress. Cells were treated with tunicamycin (TM) to induce ER stress and tauroursodeoxycholic acid (TUDCA), an ER stress inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service