Skip to Content
Merck
All Photos(2)

Documents

860102C

Avanti

16:0 Phosphatidylpropanol

1,2-dipalmitoyl-sn-glycero-3-phosphopropanol (sodium salt), chloroform

Synonym(s):

1,2-dihexadecanoyl-sn-glycero-3-phosphopropanol (sodium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H74O8PNa
CAS Number:
Molecular Weight:
712.95
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (860102C-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860102C

concentration

10 mg/mL (860102C-25mg)

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C38H75O8P.Na/c1-4-7-9-11-13-15-17-19-21-23-25-27-29-31-37(39)43-34-36(35-45-47(41,42)44-33-6-3)46-38(40)32-30-28-26-24-22-20-18-16-14-12-10-8-5-2;/h36H,4-35H2,1-3H3,(H,41,42);/q;+1/p-1/t36-;/m1./s1

InChI key

SKVWIBXHTOHOAB-SKRAEFHRSA-M

General description

Phosphatidylpropanol is a low polarity phospholipid. This phosphatidylalcohol can be produced by phospholipase D (PLD) catalyzed transphosphatidylation reaction of phosphatidylcholine (PC) and propanol.

Application

16:0 Phosphatidylpropanol or 1,2-dipalmitoyl-sn-glycero-3-phosphopropanol may be used as an internal standard in ultra-high performance liquid chromatography tandem mass spectrometry (UHPLC-MS/MS) to evaluate its matrix effect in blood samples for the validation of phosphatidylethanol (PEth). It may also be used as an internal standard for the quantification of PEth using liquid chromatography-mass spectrometry (LC-MS) in human blood.

Biochem/physiol Actions

Phosphatidylpropanol acts as a potential biomarker in phospholipase D activity assays.

Packaging

5 mL Clear Glass Sealed Ampule (860102C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Long-term stability of the alcohol consumption biomarker phosphatidylethanol in erythrocytes at- 80? C
Lakso H, et al.
Clinical mass spectrometry, 11(3), 37-41 (2019)
Mikhail Vyssotski et al.
Lipids, 44(4), 381-389 (2008-12-17)
High-performance TLC and (31)P-NMR were assessed as methods of observing the presence of numerous low polarity phospholipids: bis-phosphatidic acid (BPA), semi-lyso bis-phosphatidic acid (SLBPA), N-acyl phosphatidylethanolamine (NAPE), N-(1,1-dimethyl-3-oxo-butyl)-phosphatidylethanolamine (diacetone adduct of PE, DOBPE), N-acetyl PE, phosphatidylmethanol (PM), phosphatidylethanol (PEt), phosphatidyl-n-propanol
Siming Wang et al.
Talanta, 166, 315-320 (2017-02-19)
Phosphatidylethanol (PEth) is a special phospholipid that is only formed in the presence of ethanol, and therefore, serves as a promising biomarker for alcohol intake. In this study, a simple, rapid and precise method based on LC-MS/MS combined with ultrasound-assisted
Natalie Kummer et al.
Analytical and bioanalytical chemistry, 408(3), 825-838 (2015-11-26)
Phosphatidylethanol species (PEths) are promising biomarkers of alcohol consumption. Here, we report on the set-up, validation, and application of a novel UHPLC-ESI-MS/MS method for the quantification of PEth 16:0/18:1, PEth 18:1/18:1, and PEth 16:0/16:0 in whole blood (30 μL) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service