Skip to Content
Merck
All Photos(2)

Key Documents

830456P

Avanti

17:0 PG

1,2-diheptadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol) (sodium salt), powder

Synonym(s):

PG(17:0/17:0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H78O10PNa
CAS Number:
Molecular Weight:
773.00
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 25 mg (830456P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand 830456P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCC)OCC(O)CO

InChI

1S/C40H79O10P.Na/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-39(43)47-35-38(36-49-51(45,46)48-34-37(42)33-41)50-40(44)32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2;/h37-38,41-42H,3-36H2,1-2H3,(H,45,46);/q;+1/p-1/t37?,38-;/m1./s1

InChI key

KPXRLCBGVWDJKW-KQWGQSANSA-M

General description

Phosphatidylglycerol (PG), an anionic phospholipid is present in the intracellular membranes of mammals. 1% of the total phospholipids consists of PG.

Application

17:0 PG (1,2-diheptadecanoyl-sn-glycero-3-phospho-(1′-rac-glycerol)) has been used as an internal standard in quantifying lipids. It has also been used as a standard to examine polar metabolites and lipids profiling in human plasma using hydrophilic liquid interaction chromatography (HILIC)- fourier transform ion cyclotron resonance mass spectrometer.

Biochem/physiol Actions

The level of phosphatidylglycerol (PG) can be used to indicate the maturity of fetus lung.

Packaging

5 mL Amber Glass Screw Cap Vial (830456P-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoming Cai et al.
Scientific reports, 6, 36490-36490 (2016-11-08)
Blood plasma is the most popularly used sample matrix for metabolite profiling studies, which aim to achieve global metabolite profiling and biomarker discovery. However, most of the current studies on plasma metabolite profiling focused on either the polar metabolites or
Membrane Polar Lipids
An Introduction to Biological Membranes, 63-87 (2016)
Triacylglycerol synthesis during nitrogen stress involves the prokaryotic lipid synthesis pathway and acyl chain remodeling in the microalgae Coccomyxa subellipsoidea
Allen JW, et al.
Algal research, 10, 110-120 (2015)
Shin-Ya Morita et al.
Scientific reports, 5, 11737-11737 (2015-07-01)
Phosphatidylglycerol (PG) and cardiolipin (CL) are synthesized in mitochondria and regulate numerous biological functions. In this study, a novel fluorometric method was developed for measuring PG and CL using combinations of specific enzymes and Amplex Red. This assay quantified the
Régine Dazzoni et al.
Analytical chemistry, 92(10), 6858-6868 (2020-04-24)
The human nuclear membrane is composed of a double bilayer, the inner membrane being linked to the protein lamina network and the outer nuclear membrane continuous with the endoplasmic reticulum. Nuclear membranes can form large invaginations inside the nucleus; their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service