Skip to Content
Merck
All Photos(1)

Key Documents

810180P

Avanti

TopFluor TMR PC

Avanti Research - A Croda Brand

Synonym(s):

1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C53H82 BF2N4 O10P
Molecular Weight:
1015.02
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810180P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

General description

Tetramethylrhodamine (TMR) phosphatidylcholine (PC) is a fatty acid labeled fluorescent phospholipid. Phosphatidylcholine (PC) is the most abundant phospholipid in eukaryotes. It is the prime metabolite of glycerolipid metabolism.

Application

TopFluor TMR PC or 1-oleoyl-2-(6-((4,4-difluoro-1,3-dimethyl-5-(4-methoxyphenyl)-4-bora-3α,4α-diaza-s-indacene-2-propionyl)amino)hexanoyl)-sn-glycero-3-phosphocholine may be used for the formation of lipid membranes in microarrays.

Biochem/physiol Actions

TopFluor-labeled lipids have spectral properties similar to tetramethylrhodamine (TMR).

Packaging

5 mL Amber Glass Screw Cap Vial (810180P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rikiya Watanabe
Biochimica et biophysica acta. General subjects, 1864(2), 129330-129330 (2019-03-31)
Micro-chamber arrays enable highly sensitive and quantitative bioassays at the single-molecule level. Accordingly, they are widely used for ultra-sensitive biomedical applications, e.g., digital PCR and digital ELISA. However, the versatility of micro-chambers is generally limited to reactions in aqueous solutions
Microsystem for the single molecule analysis of membrane transport proteins
Rikiya W.
Biochimica et biophysica acta. General subjects, 1864(2), 129330-129330 (2020)
A phosphatidylinositol 4, 5-bisphosphate redistribution-based sensing mechanism initiates a phagocytosis programing.
Mu L, et al.
Nature Communications, 9(1), 4259-4259 (2018)
Rikiya Watanabe et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(12), 3066-3071 (2018-03-07)
Transmembrane protein 16F (TMEM16F) is a Ca2+-dependent phospholipid scramblase that translocates phospholipids bidirectionally between the leaflets of the plasma membrane. Phospholipid scrambling of TMEM16F causes exposure of phosphatidylserine in activated platelets to induce blood clotting and in differentiated osteoblasts to
Single-molecule analysis of phospholipid scrambling by TMEM16F
Rikiya W, et al.
Proceedings of the National Academy of Sciences of the USA, 115(12), 3066-3071 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service