Key Documents
Z422932
Sigma-Aldrich
Aldrich® pressure filter/drying funnel assemblies with removable fritted disc
funnel capacity 2,000 mL
About This Item
UNSPSC Code:
41121809
NACRES:
NB.24
material
PTFE (Bottom support)
manufacturer/tradename
Aldrich
funnel capacity
2,000 mL
pore size
coarse
capacity
2000 mL capacity
Related Categories
General description
Pressure or vacuum can be applied alternately or simultaneously for fast filtration or drying. Unique design allows the fritted disc to be removed and replaced. The bottom support is all PTFE and has a vent. A port is supplied on the top cap for an optional adjustable pressure relief valve. Large top and bottom openings are 85 mm I.D. allowing easy access to funnel contents. Complete assembly includes top cap, funnel, coarse frit, 90 mm base assembly, and 2 L bottle with GL 45 thread.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)