Key Documents
Z278521
Wilmad® coaxial insert
stem L 32 mm, reference capacity 40 μL, sample capacity 339 μL
Synonym(s):
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
UNSPSC Code:
41121705
NACRES:
SB.32
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Wilmad 520-3
stem L
32 mm
General description
Precision, all-glass construction virtually eliminates sidebands. Superior to PTFE inserts in variable-temperature experiments. Upper section of insert fits snugly into the outer NMR tube; 2 mm O.D. insert stem is held straight within 0.013 mm. Use inserts with 5 mm O.D. × 7 in. L thin-wall Precision NMR tubes only. Each insert supplied with a closing cap.
Coaxial inserts are applied in external referencing and locking, studying molecular interactions and determining magnetic susceptibility.
Outer tube must be ordered separately based on magnetic field strength.
Legal Information
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)