Key Documents
Z223727
Sigma-Aldrich
Aldrich® safety bubbler
capacity 15 mL
Synonym(s):
bubblers, safety bubblers, scientific bubblers
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
capacity
15 mL
Related Categories
General description
Built-in flash arrester bulb prevents backflow.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)