Key Documents
Z204153
Supelco
Rectangular TLC developing tanks, complete
L × H × W 17.5 cm × 16.0 cm × 8.2 cm
About This Item
UNSPSC Code:
41115711
NACRES:
NC.07
Pricing and availability is not currently available.
technique(s)
thin layer chromatography (TLC): suitable
L × H × W
17.5 cm × 16.0 cm × 8.2 cm
Footnote
TLC Developing Tanks come complete with the developing tank and lid
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Is this NMP compatible? And can it be heated?
    1 answer
    1. N-Methylpyrrolidone (NMP) is compatible with this product. It does not cause any damage or degradation to the glass surface. It is not recommended to heat the TLC-developing tanks above 100 degrees Celsius. Heating past this temperature may also cause incompatibility issues with NMP.
      Helpful?
  2. How thick are the walls of product Z204153?
    1 answer
    1. Glass thickness is 0.34 to 0.20 mm.
      Helpful?