Recommended Products
Joint
joint: ST/NS24/40
manufacturer/tradename
Ace Glass 597410
Related Categories
General description
Top of condenser is modified with Ace-Thred so that inlet is removable and replaceable. Molded spiral condensing surface. Includes body, inlet tube, and bushing.
Choose from one of the most recent versions:
Certificates of Analysis (COA)