Skip to Content
Merck
All Photos(2)

Key Documents

M59386

Sigma-Aldrich

2-Methyl-5-nitroaniline

98%

Synonym(s):

2-Amino-4-nitrotoluene, 5-Nitro-o-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)NH2
CAS Number:
Molecular Weight:
152.15
Beilstein:
879021
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

103-106 °C (lit.)

SMILES string

Cc1ccc(cc1N)[N+]([O-])=O

InChI

1S/C7H8N2O2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,8H2,1H3

InChI key

DSBIJCMXAIKKKI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anjali G Lothe et al.
Chemosphere, 242, 125117-125117 (2019-10-28)
Amino-aromatic compounds, 2-amino-4-nitrotoluene (ANT), and 2,4-diaminotoluene (DAT) are carcinogens and environmentally persistent pollutants. In this study, we investigated their degradation by natural manganese peroxidase (nMnP) derived from Phanerochaete chrysosporium and recombinant manganese peroxidase packaged in vaults (vMnP). Encapsulation of manganese
5-Nitro-ortho-toluidine.
IARC monographs on the evaluation of carcinogenic risks to humans, 48, 169-177 (1990-01-01)
H Shimizu et al.
Gut, 50(2), 266-270 (2002-01-15)
5-Nitro-o-toluidine is an aromatic nitro amino compound. While other aromatic compounds are known to damage the human liver and are registered as toxic substances, toxicity information concerning 5-nitro-o-toluidine is lacking. To investigate the hepatotoxicity of 5-nitro-o-toluidine. Of 15 workers in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service