M54503
4-(Methylthio)aniline
97%
Synonym(s):
4-(Methylmercapto)aniline, 4-Aminothioanisole
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3SC6H4NH2
CAS Number:
Molecular Weight:
139.22
Beilstein:
774506
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
97%
form
liquid
refractive index
n20/D 1.639 (lit.)
bp
272-273 °C (lit.)
density
1.119 g/mL at 25 °C (lit.)
SMILES string
CSc1ccc(N)cc1
InChI
1S/C7H9NS/c1-9-7-4-2-6(8)3-5-7/h2-5H,8H2,1H3
InChI key
YKFROQCFVXOUPW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service