DWKBDA238P
Azlon Straight-sided Beakers
graduated, polypropylene, capacity 25 mL
Synonym(s):
BDA238P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
polypropylene
feature
graduated
packaging
pkg of 5 ea
manufacturer/tradename
Azlon
storage condition
room temperature
capacity
25 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Ideal for general laboratory use and management of liquid samples
Features and Benefits
Straight sided stackable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service