Key Documents
CAD9925
Animal feeding needle, sterile, disposable, PTFE
size 15G, L × diam. 3.1 in. × 2.8 mm , ball
About This Item
UNSPSC Code:
42142609
NACRES:
NB.03
size
15G
packaging
pkg of 20 x 5 ea (5 needles per sterile Tyvek peel pouch - 20 pouches per box)
L × diam.
3.1 in. × 2.8 mm , ball
Related Categories
General description
  • PTFE tube is softer than other tubing types, very flexible, and translucent for easy confirmation of reagent delivery
  • Plastic Luer hubs and soft silicone tips
  • Non-toxic and no-pyrogenic
  • Packaged 5 needles per sterile Tyvek peel pouch - 20 pouches per box
Choose from one of the most recent versions:
Certificates of Analysis (COA)