Key Documents
BR90654
BRAND® glass beaker with spout, tall form
volume 1000 mL, with graduation
Synonym(s):
Beakers, Glass
About This Item
UNSPSC Code:
41121803
NACRES:
NB.43
material
borosilicate glass 3.3
feature
with graduation
manufacturer/tradename
BRAND 90654
volume
1000 mL
General description
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Other Notes
Browse all glass beakers
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG