BR472200
BRAND® glass staining trough with lid
soda-lime glass
Synonym(s):
Microscopy, Staining, Staining Accessories
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.13
Recommended Products
material
soda-lime glass
description
use with tray for glass staining trough; fits 10 slides
manufacturer/tradename
BRAND 472200
L × W × H
105 mm × 85 mm × 70 mm
Related Categories
General description
The BRAND® Glass Staining Trough with Lid is designed for efficient staining of up to 10 slides, making it an essential tool for laboratory applications. Constructed from durable soda-lime glass, this trough features dimensions of 76 x 26 mm, providing ample space for slide placement. The included lid helps to minimize contamination and evaporation during the staining process. For a complete staining setup, please note that the staining trough, tray, and wire handle must be ordered separately. This versatile and practical design enhances the efficiency of staining procedures in any laboratory setting.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)