BMSH3710HS
Benchmark Digital 10 x 10 inch Hotplate Magnetic Stirrer
AC/DC input 115 V
Synonym(s):
Hotplate, stirrer, stirring, magnetic Stirrers, temperature control, stirring hotplate, hot plates, ceramic top stirrer, ceramic hotplate
About This Item
material
ceramic platform
manufacturer/tradename
Benchmark Scientific H3710-HS
parameter
temp. range at Ambient +5 to 400 °C
150-1500 rpm speed
W × L
25 cm × 25 cm
Looking for similar products? Visit Product Comparison Guide
General description
Application
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service