Skip to Content
Merck
All Photos(1)

Key Documents

805106

Sigma-Aldrich

PLGA nanoparticles

200 nm average diameter

Synonym(s):

Degradex® PLGA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H4O2)x(C2H2O2)y
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

Quality Level

average diameter

200 nm

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Degradex® is made with poly(D,L-lactide-co-glycolide) with an L/G ratio of 50/50 and a Mw of 30,000.
To re-constitute:
  • remove the product from freezer
  • wait until temperature rises to the ambient temperature
  • add DI water or buffer to the container
  • Vortex and sonicate for 5 minutes to ensure the microspheres are well suspended.

Application

Plain PLGA particles for testing system compatibility in drug delivery research. Particles can be used as carriers by attaching drug or biological/chemical entities to particle surface. PLGA particles are used for controlled release, sustained release and targeted drug delivery.

Legal Information

Degradex is a registered trademark of Phosphorex, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Study on how Degradex® PLGA microspheres and nanoparticles can be used for drug delivery research and formulation development.

Study on how Degradex® PLGA microspheres and nanoparticles can be used for drug delivery research and formulation development.

Study on how Degradex® PLGA microspheres and nanoparticles can be used for drug delivery research and formulation development.

Study on how Degradex® PLGA microspheres and nanoparticles can be used for drug delivery research and formulation development.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service