Skip to Content
Merck
All Photos(1)

Documents

766453

Sigma-Aldrich

Iodo[4,5-bis(diphenylphosphino)-9,9-dimethylxanthene]copper(I)

Synonym(s):

Iodo[1,1′-(9,9-dimethyl-9H-xanthene-4,5-diyl)bis(1,1-diphenylphosphine-P)]copper(I)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H32CuIOP2
CAS Number:
Molecular Weight:
769.07
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

380-385 °C

functional group

phosphine

storage temp.

15-25°C

SMILES string

[Cu]I.CC1(C)c2cccc(P(c3ccccc3)c4ccccc4)c2Oc5c(cccc15)P(c6ccccc6)c7ccccc7

InChI

1S/C39H32OP2.Cu.HI/c1-39(2)33-25-15-27-35(41(29-17-7-3-8-18-29)30-19-9-4-10-20-30)37(33)40-38-34(39)26-16-28-36(38)42(31-21-11-5-12-22-31)32-23-13-6-14-24-32;;/h3-28H,1-2H3;;1H/q;+1;/p-1

InChI key

BEXLDSSXWBMGGT-UHFFFAOYSA-M

Application

Copper catalyst for cross coupling.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinkun Huang et al.
Journal of the American Chemical Society, 132(11), 3674-3675 (2010-02-26)
Heteroarenes are important structural moieties in many chemical industry fields. A highly efficient Pd/Cu-catalyzed C-H arylation method for a range of heterocycles has been discovered. It was found that the key to the success of this transformation is a combination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service