Skip to Content
Merck
All Photos(1)

Documents

740780

Sigma-Aldrich

Spermidine-(butyl-13C4) trihydrochloride

99 atom % 13C, 95% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2(CH2)3NH(13CH2)4NH2 • 3HCl
CAS Number:
Molecular Weight:
258.60
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.24

isotopic purity

99 atom % 13C

Quality Level

Assay

95% (CP)

form

powder

technique(s)

mass spectrometry (MS): suitable

mp

257-259 °C (lit.)

mass shift

M+4

storage temp.

−20°C

SMILES string

Cl.Cl.Cl.NCCCN[13CH2][13CH2][13CH2][13CH2]N

InChI

1S/C7H19N3.3ClH/c8-4-1-2-6-10-7-3-5-9;;;/h10H,1-9H2;3*1H/i1+1,2+1,4+1,6+1;;;

InChI key

LCNBIHVSOPXFMR-GOOBRNBHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juliette Humeau et al.
Cell death & disease, 11(6), 433-433 (2020-06-10)
The integrated stress response is characterized by the phosphorylation of eukaryotic initiation factor-2α (eIF2α) on serine 51 by one out of four specific kinases (EIF2AK1 to 4). Here we provide three series of evidence suggesting that macroautophagy (to which we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service