711268
N-tert-Butyl-O-[1-[4-(chloromethyl)phenyl]ethyl]-N-(2-methyl-1-phenylpropyl)hydroxylamine
Synonym(s):
Alkoxyamine NMP initiator
About This Item
form
solid
mp
43-47 °C
storage temp.
−20°C
SMILES string
CC(C)C(N(OC(C)c1ccc(CCl)cc1)C(C)(C)C)c2ccccc2
InChI
1S/C23H32ClNO/c1-17(2)22(21-10-8-7-9-11-21)25(23(4,5)6)26-18(3)20-14-12-19(16-24)13-15-20/h7-15,17-18,22H,16H2,1-6H3
InChI key
WZYBDTCNEIULNZ-UHFFFAOYSA-N
General description
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 3 - Skin Corr. 1B
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
>230.0 °F - closed cup
Flash Point(C)
> 110 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service