Recommended Products
Assay
97%
form
solid
mp
100-104 °C (lit.)
functional group
hydroxyl
SMILES string
COc1cc(ccc1O)C(C)O
InChI
1S/C9H12O3/c1-6(10)7-3-4-8(11)9(5-7)12-2/h3-6,10-11H,1-2H3
InChI key
BDRRAMWDUCXAKG-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service