About This Item
Recommended Products
isotopic purity
60 atom % 15N
Quality Level
form
solid
mol wt
80.64 by atom % calculation (60 atom % 15N)
bp
210 °C (lit.)
mp
169 °C (lit.)
application(s)
agriculture
mass shift
M+1
SMILES string
[O-][N+]([O-])=O.[H][15N+]([H])([H])[H]
InChI
1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3/i;1+1
InChI key
PRORZGWHZXZQMV-IEOVAKBOSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 3
Storage Class Code
5.1C - Ammonium nitrate and ammonium nitrate containing preparations
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service