536784
PAMAM dendrimer, ethylenediamine core, generation 5.5 solution
5 wt. % in methanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[NH2(CH2)2NH2]:(G=5.5);dendri PAMAM(NHCH2CH2COONa)256
CAS Number:
Molecular Weight:
52900.21
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Recommended Products
description
sodium carboxylate surface groups
form
liquid
mol wt
generation 5.5
feature
core type ethylenediamine core (2-carbon core)
concentration
5 wt. % in methanol
no. Surface Groups
256
refractive index
n20/D 1.337
density
0.8 g/mL at 25 °C
storage temp.
2-8°C
Related Categories
Physical form
Contains 256 surface carboxylate groups.
Legal Information
Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1
Target Organs
Eyes
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
51.8 °F - closed cup
Flash Point(C)
11 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Maria Dąbkowska et al.
Journal of nanobiotechnology, 18(1), 120-120 (2020-09-02)
Brain-derived neurotrophic factor (BDNF) is essential for the development and function of human neurons, therefore it is a promising target for neurodegenerative disorders treatment. Here, we studied BDNF-based electrostatic complex with dendrimer nanoparticles encapsulated in polyethylene glycol (PEG) in neurotoxin-treated
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service