Skip to Content
Merck
All Photos(1)

Key Documents

491640

Sigma-Aldrich

Acetic acid-17O2

20 atom % 17O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C17O2H
CAS Number:
Molecular Weight:
62.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

20 atom % 17O

Quality Level

mol wt

60.46 by atom % calculation (10 atom % 17O)

refractive index

n20/D 1.3715 (lit.)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

density

1.056 g/mL at 25 °C

mass shift

M+2

SMILES string

CC([17OH])=[17O]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i3+1,4+1

InChI key

QTBSBXVTEAMEQO-CQDYUVAPSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service