About This Item
Recommended Products
isotopic purity
99 atom % D
Quality Level
Assay
99% (CP)
technique(s)
mass spectrometry (MS): suitable
refractive index
n20/D 1.5265 (lit.)
bp
247 °C/745 mmHg (lit.)
density
1.029 g/mL at 25 °C (lit.)
application(s)
environmental
mass shift
M+3
storage temp.
−20°C
SMILES string
[2H]C([2H])([2H])N1CCCC1c2cccnc2
InChI
1S/C10H14N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2,4,6,8,10H,3,5,7H2,1H3/i1D3
InChI key
SNICXCGAKADSCV-FIBGUPNXSA-N
Related Categories
Packaging
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service