Skip to Content
Merck
All Photos(2)

Documents

472093

Sigma-Aldrich

Ethylenediamine, polymer-bound

100-200 mesh, extent of labeling: 4.0-5.7 mmol/g loading, 1 % cross-linked

Synonym(s):

Ethylenediamine on polystyrene, Ethylenediaminomethyl polystyrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

form

solid

crosslinking

1 % cross-linked

reaction suitability

reaction type: solution phase peptide synthesis

extent of labeling

4.0-5.7 mmol/g loading

particle size

100-200 mesh

functional group

amine

SMILES string

NCCN.C=Cc1ccccc1.C=Cc2ccc(C=C)cc2

InChI

1S/C6H20N6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-6H,1-2,7-12H2

InChI key

PIJGCIFQPQGTTG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Scavenges aldehydes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flynn, D.L.
Journal of the American Chemical Society, 119, 4874-4874 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service