462330
Poly(1-decene), hydrogenated
Mixture of trimer and tetramer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
[CH2CH[(CH2)7CH3]]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
refractive index
n20/D 1.455 (lit.)
viscosity
20 cSt(40 °C)(lit.)
bp
>316 °C (lit.)
density
0.818 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCC=C
InChI
1S/C10H20/c1-3-5-7-9-10-8-6-4-2/h3H,1,4-10H2,2H3
InChI key
AFFLGGQVNFXPEV-UHFFFAOYSA-N
Related Categories
General description
Hydrogenated poly(1-decene) is prepared by the hydrogenation of the mixture of isoparaffinic molecules (trimer and tetramer).
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Compendium of food additive specifications.
FAO JECFA Monographs (2006)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service