Skip to Content
Merck
All Photos(2)

Key Documents

423548

Sigma-Aldrich

N-Isopropylmethacrylamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=C(CH3)CONHCH(CH3)2
CAS Number:
Molecular Weight:
127.18
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

89-91 °C (lit.)

SMILES string

CC(C)NC(=O)C(C)=C

InChI

1S/C7H13NO/c1-5(2)7(9)8-6(3)4/h6H,1H2,2-4H3,(H,8,9)

InChI key

YQIGLEFUZMIVHU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adriana M Mihut et al.
Science advances, 3(9), e1700321-e1700321 (2017-09-21)
We have seen a considerable effort in colloid sciences to copy Nature's successful strategies to fabricate complex functional structures through self-assembly. This includes attempts to design colloidal building blocks and their intermolecular interactions, such as creating the colloidal analogs of
Viktor Sabadasch et al.
Soft matter, 16(23), 5422-5430 (2020-06-04)
Responsive core-shell microgels are promising systems for a stabilization of Pd nanoparticles and control of their catalytic activity. Here, poly-N-n-propylacrylamide (PNNPAM) was copolymerized with methacrylic acid to yield microgel core particles, which were subsequently coated with an additional, acid-free poly-N-isopropylmethacrylamide
S Brosel-Oliu et al.
Lab on a chip, 19(8), 1436-1447 (2019-03-19)
The presence of antimicrobial contaminants like antibiotics in the environment is a major concern because they promote the emergence and the spread of multidrug resistant bacteria. Since the conventional systems for the determination of bacterial susceptibility to antibiotics rely on
Timo Brändel et al.
Polymers, 11(8) (2019-08-03)
The present study focuses on the development of multiresponsive core-shell microgels and the manipulation of their swelling properties by copolymerization of different acrylamides-especially N-isopropylacrylamide (NIPAM), N-isopropylmethacrylamide (NIPMAM), and NNPAM-and acrylic acid. We use atomic force microscopy for the dry-state characterization
Marian Cors et al.
Polymers, 11(4) (2019-04-10)
The effect of deuteration on the volume phase transition (VPT) temperature of poly (N-isopropylmethacrylamide) (pNIPMAM) microgels in aqueous suspension is determined via IR spectroscopy and size measurements by photon correlation spectroscopy (PCS). We study the effect of a hydrogenated and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service