Skip to Content
Merck
All Photos(1)

Documents

275719

Sigma-Aldrich

4′-Ethoxyacetophenone

98%

Synonym(s):

4′-ethoxy acetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H4COCH3
CAS Number:
Molecular Weight:
164.20
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

solid

bp

268-269 °C/758 mmHg (lit.)

mp

37-39 °C (lit.)

SMILES string

CCOc1ccc(cc1)C(C)=O

InChI

1S/C10H12O2/c1-3-12-10-6-4-9(5-7-10)8(2)11/h4-7H,3H2,1-2H3

InChI key

YJFNFQHMQJCPRG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service