Skip to Content
Merck
All Photos(2)

Key Documents

259101

Sigma-Aldrich

Bis(2-diphenylphosphinoethyl)phenylphosphine

97%

Synonym(s):

Phenylbis(diphenylphosphinoethyl)phosphine, Triphos

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5P[CH2CH2P(C6H5)2]2
CAS Number:
Molecular Weight:
534.55
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

mp

130-132 °C (lit.)

functional group

phosphine

SMILES string

C(CP(c1ccccc1)c2ccccc2)P(CCP(c3ccccc3)c4ccccc4)c5ccccc5

InChI

1S/C34H33P3/c1-6-16-30(17-7-1)35(26-28-36(31-18-8-2-9-19-31)32-20-10-3-11-21-32)27-29-37(33-22-12-4-13-23-33)34-24-14-5-15-25-34/h1-25H,26-29H2

InChI key

AXVOAMVQOCBPQT-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daniel T Walters et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(15), 3849-3857 (2018-12-24)
Two new molecular boxes, the mono-bromo box [Au6 (Triphos)4 Br](SbF6 )5 ⋅6(CH2 Cl2 ), 4 mB, and the dibromo box, [Au6 (Triphos)4 Br2 ⋅H2 O](SbF6 )4 ⋅4(CH2 Cl2 ), 5 dB, have been prepared in crystalline form. Although constructed from non-luminescent components

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service