Skip to Content
Merck
All Photos(1)

Documents

182079

Sigma-Aldrich

Poly(2-ethylhexyl methacrylate) solution

average Mw ~100,000 by GPC, in toluene

Synonym(s):

2-Ethylhexyl methacrylate homopolymer, Poly(ethylhexyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H22O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor density

1.2 (37 °C, vs air)

form

liquid

mol wt

average Mw ~100,000 by GPC

concentration

in toluene

weight

Unit weight does not include weight of solvent.

transition temp

Tg −10 °C

density

0.908 g/mL at 25 °C

SMILES string

CCCCC(CC)COC(=O)C(C)=C

InChI

1S/C12H22O2/c1-5-7-8-11(6-2)9-14-12(13)10(3)4/h11H,3,5-9H2,1-2,4H3

InChI key

WDQMWEYDKDCEHT-UHFFFAOYSA-N

General description

PEHMA exhibits biomimetic properties. It finds potential application as a flexible hydrogel for ophthalmologic applications.

Application

Poly(2-ethylhexyl methacrylate) (PEHMA) may be used to synthesize binary polymeric blends with poly(butyl methacrylate). PEHMA may be utilized in the preparation of PEHMA /carbon black composites for gas sensing. PHEMA-antioxidant conjugate (Quercetin (Q) ) may be prepared for ophthalmologic uses.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poly (2-hydroxyethyl methacrylate)-quercetin Conjugate as Biomaterial in Ophthalmology: An ?ab initio? Study.
Curcio M, et al.
Journal of Functional Biomaterials, 2(1), 1-17 (2011)
Carbon black filled poly (2-ethylhexyl methacrylate) as a candidate for gas sensing material.
Dong X M, et al.
Journal of Materials Science Letters, 22(15), 1057-1059 (2003)
An energy tranfer study of the interface thickness in blends of poly (butyl methacrylate) and poly (2-ethylhexyl methacrylate).
Farinha J P S, et al.
Macromolecules, 33(16), 5863-5873 (2000)

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service