Skip to Content
Merck
All Photos(2)

Key Documents

177792

Sigma-Aldrich

Diethyl dithiophosphate ammonium salt

95%

Synonym(s):

Ammonium O,O-diethyl dithiophosphate, Ammonium O,O-diethyl phosphorodithioate, Ammonium O,O-diethyl phosphorodithioate(1-), Ammonium O,O′-diethyldithiophosphate, Ammonium di-O-ethyl dithiophosphate, Ammonium diethyldithiophosphate, O,O-Diethyl ammonium phosphorodithioate, O,O-Diethylphosphorodithioic acid ammonium salt, S-Ammonium O,O′-diethyl phosphorodithioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(S)SNH4
CAS Number:
Molecular Weight:
203.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

164-166 °C (lit.)

SMILES string

N.CCOP(S)(=S)OCC

InChI

1S/C4H11O2PS2.H3N/c1-3-5-7(8,9)6-4-2;/h3-4H2,1-2H3,(H,8,9);1H3

InChI key

HFRHTRKMBOQLLL-UHFFFAOYSA-N

Application

Diethyl dithiophosphate ammonium salt was used in the extraction of arsenic.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verónica Arancibia et al.
Talanta, 68(5), 1567-1573 (2008-10-31)
The separation of arsenic based on in situ chelation with ammonium diethyl dithiophosphate (ADDTP) has been carried out using methanol-modified supercritical CO(2). Aliquots of extract were added to an electroanalytical cell and arsenic was determined by square wave cathodic stripping
Jeferson Schneider Carletto et al.
Analytica chimica acta, 638(1), 45-50 (2009-03-21)
A hollow fiber renewal liquid membrane (HFRLM) extraction method to determine cadmium (II) in water samples using Flame Atomic Absorption Spectrometry (FAAS) was developed. Ammonium O,O-diethyl dithiophosphate (DDTP) was used to complex cadmium (II) in an acid medium to obtain
César Ricardo Teixeira Tarley et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 21(11), 1337-1342 (2005-12-02)
The present paper describes the on-line coupling of a flow-injection system to a new technique, thermospray flame furnace-AAS (TS-FF-AAS), for the preconcentration and determination of copper in water samples. Copper was preconcentrated onto polyurethane foam (PUF) complexed with ammonium O,O-diethyldithiophosphate
Li-Shing Huang et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 22(10), 1375-1378 (2006-10-14)
Inductively coupled plasma-mass spectrometry (ICP-MS) incorporated with an on-line preconcentration system was used to determine trace amounts of ten metals including Ni, Cu, Zn, Rh, Ag, Cd, In, Au, Tl, and Pb in aqueous solutions and seawater. These metals, which
K Borg et al.
Archives of toxicology, 62(5), 387-391 (1988-01-01)
Brown trout, Salmo trutta, were exposed to water containing 0.1 microgram/l 203Hg2+, alone or with potassium ethylxanthate (PEX), sodium isopropylxanthate (SIX), sodium diethyldithiophosphate (SEP), sodium diisopropyldithiophosphate (SIP), sodium dimethyldithiocarbamate (SMC), sodium diethyldithiocarbamate (SEC) or sodium pyridinethione (SPyr), respectively. After 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service