Skip to Content
Merck
All Photos(3)

Key Documents

148008

Sigma-Aldrich

Methyl isobutyrate

99%

Synonym(s):

2-Methylpropanoic acid methyl ester, Methyl 2,2-dimethylacetate, Methyl 2-methylpropanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCOOCH3
CAS Number:
Molecular Weight:
102.13
Beilstein:
1740720
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.384 (lit.)

bp

90 °C (lit.)

solubility

alcohol: miscible
diethyl ether: miscible
water: slightly soluble

density

0.891 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)C(C)C

InChI

1S/C5H10O2/c1-4(2)5(6)7-3/h4H,1-3H3

InChI key

BHIWKHZACMWKOJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Radiofrequency-induced plasmas of methyl isobutyrate has been studied using mass spectrometry.

Application

Methyl isobutyrate was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Penetrant diffusion in poly (methyl methacrylate) near Tg: dependence on temperature and polymer weight fraction.
Tonge MP, et al.
Polymer, 41(10), 3659-3670 (2000)
Mass spectrometry of and deposition-rate measurements from radiofrequency-induced plasmas of methyl isobutyrate, methyl methacrylate and n-butyl methacrylate.
O'Toole L, et al.
J. Chem. Soc., Faraday, 91(9), 1363-1370 (1995)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service