All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H13NO
CAS Number:
Molecular Weight:
175.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
form
liquid
refractive index
n20/D 1.552 (lit.)
density
1.095 g/mL at 25 °C (lit.)
functional group
phenyl
SMILES string
O=C1CCCN1Cc2ccccc2
InChI
1S/C11H13NO/c13-11-7-4-8-12(11)9-10-5-2-1-3-6-10/h1-3,5-6H,4,7-9H2
InChI key
LVUQCTGSDJLWCE-UHFFFAOYSA-N
General description
1-Benzyl-2-pyrrolidinone undergoes reduction in THF with lithium N,N-dialkylaminoborohydrides to yield N-benzylpyrrolidine.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J M Flaniken et al.
Organic letters, 1(5), 799-801 (2005-08-25)
[reaction: see text] Various five- and six-membered N-alkyl lactams were reduced to the corresponding cyclic amines using lithium N,N-dialkylaminoborohydrides (LAB). Most of the reductions were essentially complete after refluxing in THF for 2 h. The cyclic amine products were easily
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service