Merck
  • Development of dinuclear vanadium catalysts for enantioselective coupling of 2-naphthols via a dual activation mechanism.
Development of dinuclear vanadium catalysts for enantioselective coupling of 2-naphthols via a dual activation mechanism.
Chemical & pharmaceutical bulletin (2009-11-03)
Shinobu Takizawa
ABSTRACT
This review describes our recent efforts in the development of chiral dinuclear vanadium complexes which work as dual activation catalysts for oxidative coupling of 2-naphthols. A chiral dinuclear vanadium(IV) complex (R(a),S,S)-1a possessing (S)-tert-leucine moieties at the 3,3'-positions of the (R)-binaphthyl skeleton was developed, and found to promote oxidative coupling of 2-naphthol to afford (S)-1,1'-bi-2-naphthol (BINOL) with 91% ee. To verify the dual activation mechanism, mononuclear vanadium(IV) complex (S)-10 was also prepared. Kinetic analysis revealed that the reaction rate of oxidative coupling of 2-naphthol promoted by (R(a),S,S)-1a is 48.3 times faster than that of (S)-10. In the coupling reaction, the two vanadium metals in the chiral complex activate two molecules of 2-naphthol simultaneously achieving a high reaction rate with high enantiocontrol. Since the dinuclear vanadium(IV) complex was found to be readily oxidized to afford a corresponding vanadium(V) species during preparation in air, a new synthetic procedure using VOCl3 and a convenient one-pot procedure using VOSO4 under O2 have been applied towards dinuclear vanadium(V) complexes (R(a),S,S)-2. The coupling of 2-naphthol was catalyzed by (R(a),S,S)-2 with reaction rates enhanced 2.3 times faster than that of (R(a),S,S)-1a, which is likely a precatalyst. To the best of our knowledge, (R(a),S,S)-1a, 2, and 3 show considerably higher catalytic activity than previously reported vanadium complexes for oxidative coupling of 2-naphthols.
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Vanadium, wire reel, 2m, diameter 0.5mm, as drawn, 99.8%
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 5m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 5m, diameter 0.5mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 1.0μm, specific density 611μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 1m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.25mm, hard, 99.8%
Vanadium, wire reel, 1m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, microfoil, disks, 10mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, microfoil, disks, 25mm, thinness 0.1μm, specific density 60.8μg/cm2, permanent mylar 3.5μm support, 99.8+%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%