Merck
  • Controversies in endocrinology: On the need for universal thyroid screening in pregnant women.
Controversies in endocrinology: On the need for universal thyroid screening in pregnant women.
European journal of endocrinology (2013-10-17)
Lluís Vila, Inés Velasco, Stella González, Francisco Morales, Emilia Sánchez, Sara Torrejón, Berta Soldevila, Alex Stagnaro-Green, Manuel Puig-Domingo
ABSTRACT
There is a well-known controversy among scientific societies regarding the recommendation to screen for thyroid dysfunction (TD) during pregnancy. Although several studies have shown an association between maternal subclinical hypothyroidism and/or hypothyroxinemia with obstetric problems and/or neurocognitive impairment in the offspring, there is only limited evidence on the possible positive effects of thyroxine (T4) treatment in such cases. Despite the scarcity of this evidence, there is a widespread agreement among clinicians on the need for treatment of clinical hypothyroidism during pregnancy and the risks that could arise due to therapeutic abstention. As maternal TD is a quite prevalent condition, easily diagnosed and for which an effective and safe treatment is available, some scientific societies have proposed to assess thyroid function during the first trimester of pregnancy and ideally before week 10 of gestational age. Given the physiologic changes of thyroid function during pregnancy, hormone assessment should be performed using trimester-specific reference values ideally based on locally generated data as geographic variations have been detected. Screening of TD should be based on an initial determination of TSH performed early during the first trimester and only if abnormal should it be followed by either a free or total T4 measurement. Furthermore, adequate iodine supplementation during pregnancy is critical and if feasible it should be initiated before the woman attempts to conceive.
Product Description
Supelco
L-Thyroxine (T4) solution, 100 μg/mL in methanol with 0.1N NH3, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Thyrotropic hormone from human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)