Merck
  • Simultaneous determination of arsenic and selenium in biological samples by HG-AFS.
Simultaneous determination of arsenic and selenium in biological samples by HG-AFS.
Analytical and bioanalytical chemistry (2005-06-02)
Hanwen Sun, Zhanfeng Liu, Wenjuan Wu, Liqing Li, Hongmei Shi
ABSTRACT
A new method is proposed for simultaneous determination of traces of arsenic (As) and selenium (Se) in biological samples by hydride-generation double-channel non-dispersive atomic-fluorescence spectrometry (HG-AFS) from tartaric acid media. The effects of analytical conditions on fluorescence signal intensity were investigated and optimized. Interferences from coexisting ions were evaluated. Under optimum conditions linear response ranges above 20 microg L(-1) for As and 32 microg L(-1) for Se were obtained with detection limits of 0.13 and 0.12 microg L(-1), respectively. The precision for eleven-fold determination of As at the 4 microg L(-1) level and of Se at the 8 microg L(-1) level were 2.7 and 1.9% (RSD), respectively. Recoveries of 92.5-95.5% for As and 101.2-108.4% for Se were obtained for four biological samples and two certified biological reference materials. The proposed method has the advantages of simple operation, high sensitivity, and high efficiency; it was successfully used for simultaneous determination of As and Se in biological samples.
Sigma-Aldrich
Potassium borohydride, 99.9% trace metals basis
Sigma-Aldrich
Potassium borohydride, purum, ≥97.0% (RT)
Sigma-Aldrich
Potassium borohydride, ≥97%