Merck
  • A thermoreversible hydrogel as a biosynthetic bandage for corneal wound repair.
A thermoreversible hydrogel as a biosynthetic bandage for corneal wound repair.
Biomaterials (2007-11-03)
Chayanin Pratoomsoot, Hidetoshi Tanioka, Kuniko Hori, Satoshi Kawasaki, Shigeru Kinoshita, Patrick J Tighe, Harminder Dua, Kevin M Shakesheff, Felicity Rosamari A J Rose
ABSTRACT
Ocular trauma and disorders that lead to corneal blindness account for over 2 million new cases of monocular blindness every year. A popular ocular surface reconstruction therapy, amniotic membrane transplantation, has been shown to aid corneal wound repair. However, the success rates of the procedure are variable. Here, we proposed to bioengineer a novel synthetic material that would serve as a biomimetic corneal bandage. The PLGA-PEG-PLGA triblock copolymer was synthesised via ring-opening polymerisation. Thermoreversible gelation behaviour was investigated at different polymer concentrations (23%, 30%, 35%, 40%, 45%, w/v) at temperatures ranging between 5 and 60 degrees C. Viscoelastic properties were studied in dynamic mechanical analysis with 1 degrees C/min temperature ramp. Cryo-SEM revealed a porous hydrogel with interconnecting networks. No adverse cytotoxicity was observed with an in vitro scratch-wound assay and in in vivo biocompatibility tests. We have demonstrated that the PLGA-PEG-PLGA hydrogel possessed a suitable gelling profile and, for the first time, the biocompatibility properties for this application as a potential bandage for corneal wound repair.
Sigma-Aldrich
Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide), average Mn (1,700-1,500-1,700), lactide:glycolide (95:5)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(lactide-co-glycolide)-b-poly(ethylene glycol)-b-poly(lactide-co-glycolide), average Mn (1,600-1,500-1,600), lactide:glycolide 75:25