Skip to Content
BW
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
DURAN
®
culture tubes
Products
Product Category
tubes (6)
vials (3)
Feature
Manufacturer
Capacity (mL)
Diameter (mm)
Cap
Color
Package Size
Sterility
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
DWK261352254
DURAN
®
culture tubes,
glass tube diam. × H 16 mm × 150 mm, GL 18 thread, tube capacity 20 mL, with cap, borosilicate glass 3.3 tube
Expand
View Pricing
DWK261351258
DURAN
®
culture tubes,
GL 14 thread, tube capacity 9 mL, with cap, borosilicate glass 3.3 tube
Expand
View Pricing
DWK261352451
DURAN
®
culture tubes,
GL 18 thread, tube capacity 34 mL, with cap, borosilicate glass 3.3 tube
Expand
View Pricing
Z620262
DURAN
®
culture tubes,
H × diam. 180 mm × 18 mm
Expand
View Pricing
Z620246
DURAN
®
culture tubes,
H × diam. 100 mm × 12 mm
Expand
View Pricing
Z620254
DURAN
®
culture tubes,
H × diam. 160 mm × 16 mm
Expand
View Pricing