
Boldine

Synonym(s):
2,9-Dihydroxy-1,10-dimethoxyaporphine
Empirical Formula (Hill Notation):
C19H21NO4
CAS Number:
Molecular Weight:
327.37
Products
Brand
Biological Source
Formula Weight
Manufacturer
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing