SAYB08
Saybolt Colour Reference Standard
ASTM D 6045, D 156 (ASTM colour -10)
About This Item
Recommended Products
grade
certified reference material
Agency
according to ASTM® D 156
according to ASTM® D 6045
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Paragon Scientific Ltd
application(s)
cleaning products
coatings
cosmetics
environmental
flavors and fragrances
food and beverages
glass & ceramic
personal care
pharmaceutical
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Full traceability to international standards with maximum data accuracy
- Colour values compliant to ASTM D 6045 and D 156 colour scales
- 500-mL pack size available with safety data sheet
- 18-months shelf life
- Tamper-evident security packaging
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
168.8 °F
Flash Point(C)
76 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service