SAAR5766040
Silver sulfate
univAR™
Synonym(s):
Sulfuric acid disilver(I) salt
About This Item
product line
univAR™
packaging
pkg of 100 g (SAAR5766040DC)
mp
652 °C (lit.)
storage temp.
2-30°C
SMILES string
[Ag+].[Ag+].[O-]S([O-])(=O)=O
InChI
1S/2Ag.H2O4S/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2
InChI key
YPNVIBVEFVRZPJ-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service