Skip to Content
Merck
All Photos(1)

Key Documents

SAAR3745000

Kovacs Indole Reagent

indicator solution, uniLAB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116100

Product Name

Kovacs Indole Reagent,

product line

uniLAB

packaging

pkg of 100 mL (SAAR3745000JT)

storage temp.

2-8°C

Application

For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 3 - Met. Corr. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

110.3 °F - closed cup

Flash Point(C)

43.5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service