Skip to Content
Merck
All Photos(1)

Key Documents

SAAR1123030

Ammonium fluoride

uniLAB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
H4FN
CAS Number:
Molecular Weight:
37.04
UNSPSC Code:
12352302

product line

uniLAB

packaging

poly bottle of 1 × 500 g (SAAR1123030EM)

density

1.01 g/cm3

storage temp.

2-30°C

SMILES string

F.N

InChI key

LDDQLRUQCUTJBB-UHFFFAOYSA-N

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service