Skip to Content
Merck
All Photos(1)

Documents

SAAR1122020

Ammonia solution

25%, univAR

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12142100

product line

univAR

packaging

glass bottle of 4 × 2.5 L (SAAR1122020LC)

concentration

25%

storage temp.

2-30°C

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Analysed reagent
For laboratory use only

Legal Information

univAR is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service