Skip to Content
Merck
All Photos(2)

Key Documents

23991

Supelco

Molecular Sieve 5A Moisture Trap

volume 750 cc, 1/4 in. fitting

Synonym(s):

BIG MOISTURE TRAP, 0.75 LITER, 1/4IN FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
Pricing and availability is not currently available.

volume

750 cc

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

Molecular Sieve 5A efficiently removes moisture and heavy hydrocarbons from compressed air, electrolytically produced hydrogen, house nitrogen, or other gases with high moisture or hydrocarbon content.

Dimensions:
  • 200 cc Trap: 26 1/4 in. (67 cm) long × 1 in. (2.5 cm) diameter
  • 750 cc Trap: 18 in. (45.7 cm) long × 2 3/8 in. (6 cm) diameter
  • S-Trap: 7 1/2 in. (19 cm) long x 5 9/16 in. (14.1 cm) wide x 1/2 in. (13 mm) diameter [total bed length: 19 3/4 in. (50.2 cm); the extended bed length ensures prolonged contact between the gas and the adsorbent and provides greater working capacity]

Application

Molecular Sieve 5A moisture trap may be used for the removal of hydrocarbons from air.[1] Furthermore, it may also be used in gas chromatography for the measurement of atmospheric nitrous oxide.[2]

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C J Dillard et al.
Lipids, 12(1), 109-114 (1977-01-01)
An analytical method for the measurement of hydrocarbon gases in the breath of rats is described. The method was used to follow the expiration in rat breath of in vivo formed scission products of hydroperoxides. The major products are pentane
Measurement of atmospheric nitrous oxide using a molecular sieve 5A trap and gas chromatography.
Miles LD.
Analytical Chemistry, 43 (8), 1113-1115 (1971)

Questions

1–3 of 3 Questions  
  1. Hi, We just purchased the Molecular Sieve 5A. What flow rate could we expect to get with an inlet pressure of 200 psi? If we connected another Sieve 5a in parallel would we double the flow rate? Thanks Vic

    1 answer
    1. The 200 cc Molecular Sieve 5A Trap is rated for a maximum flow rate of 2,000 cc/min at an inlet pressure of 250 psig/17.2 bar. The 750 cc Molecular Sieve 5A Trap is rated for a maximum flow rate of 10,000 cc/min at an inlet pressure of 250 psig/17.2 bar. In theory, if the same inlet pressure can be maintained for two traps run in parallel, the effective flow rate measured from the combined outlets would double. In practice, this flow rate may not be exactly doubled due to some uncontrollable variables between a single trap and multi-trap system.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. How do I refill Product No. 23991, Molecular Sieve 5A Water Vapor Trap?

    1 answer
    1. Product No. 23991 at one end has a two piece fitting.  Remove the 1/8 in NPT fitting and empty contents into an appropriate container for proper disposal.  Clean and replace any Teflon sealing material. Refill the trap with four of Product No. 20298.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service