Skip to Content
Merck
All Photos(6)

Key Documents

ZRB2650

Sigma-Aldrich

Anti-Cas9 Antibody, clone 1C12-H1 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CRISPR-associated endonuclease Cas9/Csn1, EC:3.1.-.-, SpCas9, SpyCas9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C12-H1, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 158 kDa
observed mol wt ~158 kDa

purified by

using Protein A

species reactivity

all

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgGκ

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C12-H1 is a ZooMAb® rabbit recombinant monoclonal antibody that detects Cas9. It targets an epitope within 24 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the C-terminal part of human Cas9.

Application

Quality Control Testing

Evaluated by Western Blotting in lysate from Cas9 transfected HEK-293 cells.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Cas9 in lysate from HEK-293 cells transfected with Cas9.

Tested Applications

Flow Cytometry Analysis: 1 µg from a representative lot detected Cas9 in one million NIH/3T3 cells transfected with Cas9.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected Cas9 in NIH/3T3 cells transfected with Cas9.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

CRISPR-associated endonuclease Cas9/Csn1 (UniProt: Q99ZW2; also known as EC:3.1.-.-, SpCas9, SpyCas9) is encoded by the cas9 (also known as csn1, SPy_1046) gene in Streptococcus pyogenes. CRISPR-associated endonuclease Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria. Cas9 plays a crucial role in the defense mechanism against against mobile genetic elements (viruses, transposable elements and conjugative plasmids) by cleaving foreign DNA. Cas9 has a bilobed architecture, with a recognition lobe that binds RNA and a nuclease lobe that contains the HNH and RuvC-like nuclease domains. These domains are responsible for cleaving the DNA strands complementary to the guide RNA and the opposite strand, respectively. The enzyme requires a guide RNA that is complementary to the target DNA sequence and a protospacer adjacent motif (PAM) sequence downstream of the target for efficient binding and cleavage. The primary function of Cas9 is to introduce double-stranded breaks at specific locations in the DNA, which are then repaired by the cell′s native repair mechanisms. This feature is harnessed in genome editing to introduce deletions, insertions, or replacements in the DNA sequence. Cas9 application in genome editing has significant therapeutic potential. By targeting and modifying disease-associated genes, Cas9 has been explored as a tool to treat genetic disorders, including sickle cell disease, cystic fibrosis, and certain types of cancer. However, the off-target effects and ethical considerations of genome editing are areas of active investigation and debate. The precise and controlled use of Cas9 remains a critical factor in translating its capabilities into safe and effective therapies.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service