Skip to Content
Merck
All Photos(7)

Key Documents

ZRB1595

Sigma-Aldrich

Anti-IKKβ Antibody, clone 2N22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Inhibitor of nuclear factor kappa-B kinase subunit beta;EC:2.7.11.10;I-kappa-B-kinase beta;IKK-B;IKK-beta;IkBKB;I-kappa-B kinase 2;IKK2;Nuclear factor NF-kappa-B inhibitor kinase beta;NFKBIKB;Serine/threonine protein kinase IKBKB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2N22, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 86.56 kDa
observed mol wt ~90 kDa

purified by

using Protein A

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

bovine, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2N22 is a Rabbit recombinant monoclonal antibody that specifically detects IKKb. It targets an epitope within 24 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half of human IKKb.

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected IKKβ in A549 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected IKKβ in HepG2, L6, and NIH3T3 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound IKKb with a KD of 7.9 x 10-8 in an affinity binding assay.

Enzyme Immunoassay (ELISA) Analysis: Various dilutions of this antibody detected IKKβ in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Inhibitor of nuclear factor kappa-B kinase subunit beta (UniProt: O14920; also known as EC:2.7.11.10, I-kappa-B-kinase beta, IKK-B, IKK-beta, IkBKB, I-kappa-B kinase 2, IKK2, Nuclear factor NF-kappa-B inhibitor kinase beta, NFKBIKB, Serine/threonine protein kinase IKBKB) is encoded by the IKBKB (also known as IKKB) gene (Gene ID: 3551) in human. The IKK complex is composed of three subunits, IKKa , IKKb, and IKKg (NEMO). While IKKa and IKKb are highly similar catalytic subunits and can phosphorylate IkB, IKKg is the regulatory subunit. IKKb is a serine kinase that plays an essential role in the NF-kB signaling pathway. It acts as a part of the canonical IKK complex in the conventional pathway of NF-kB activation. Its protein kinase domain is localized in amino acids 15-300. It phosphorylates inhibitors of NF-kB (IkB) on two critical serine residues that allow polyubiquitination of these inhibitors and their subsequent degradation by the proteasome. This allows free NF-kB to translocate into the nucleus and activate the transcription of genes involved in immune response, growth control, and protection against apoptosis. Upon cytokine stimulation, IKKb is phosphorylated on serine 177 and serine 181 by MEKK1 and/or MAP3K14/NIK as well as TBK1 and protein kinase Cz that enhances its activity. Once activated, it undergoes autophosphorylation on the C-terminal serine cluster, which reduces its activity and prevents prolonged activation of the inflammatory response. Mutations in IKBKB gene are known to cause life threatening immunodeficiency that develops in in infancy and increases susceptibility to bacterial, fungal, and viral infections. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Li, ZW., et al. (1999). J. Exp. Med. 189(11); 1839-1845).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Learn about the hallmarks of cancer and find reliable ZooMAb® recombinant antibodies to study them, conveniently organized by cancer hallmark.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service