Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1194

Sigma-Aldrich

Anti-KLF5 Antibody, clone 1J12 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J12, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~50.79 kDa
observed mol wt ~60 kDa

species reactivity

human, mouse

species reactivity (predicted by homology)

monkey, bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J12 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Krueppel-like factor 5 (KLF5). It targets an epitope within 22 amino acids from the C-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human Krueppel-like factor 6 (KLF5).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected KLF5 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected KLF5 in NIH3T3 cell lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected KLF5 in human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound KLF5 peptide with a KD of 2.0 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected KLF5 in PC3 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-KLF5, clone 1J12 ZooMAb®, Cat. No. ZRB1194, is a recombinant Rabbit monoclonal antibody that specifically targets KLF5 and is tested for use in in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Krueppel-like factor 5 (UniProt: Q13887; also known as Basic transcription element-binding protein 2, BTE-binding protein 2, Colon krueppel-like factor, GC-box-binding protein 2, Intestinal-enriched krueppel-like factor, Transcription factor BTEB2) is encoded by the KLF5 (also known as BTEB2, CKLF, IKLF) gene (Gene ID: 688) in human. KLF5 is a member of the krueppel C2H2-type zinc-finger protein family that is expressed in testis and placenta. It serves as a transcription factor that binds to GC box promoter elements and activates the transcription of several genes. It contributes to the regulation of various cellular processes, including cell proliferation, differentiation, angiogenesis, and migration. KLF5 is reported to function as an oncogenic transcription factor in several cancer types, including breast, bladder, and intestinal cancers. Its expression can be induced by various oncogenic and pro-inflammatory factors, including TNF- , lipopolysaccharides, and interleukin-1β. Co-expression of KLF5 and TNFRSF11 correlates significantly with tumorigenesis in cervical tissues where it promotes cell proliferation, migration, and invasiveness in a manner dependent partly on TNFRSF11 expression. TNF- induced KLF5 is shown to positively regulate TNFRSF11 expression by binding to the TNFRSF11 promoter. KLF5 expression is reported to be regulated by estrogen and progesterone in breast cancer cells where estrogen facilitates KLF5 degradation and progesterone induces its expression. Loss of KLF5 in uterine tissue is reported to cause infertility due to defective implantation. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Ma, D., et al. (2017). Sci. Rep. 7; 15683; Sun, X., et al. (2012). Proc. Natl. Acad. Sci. USA. 109(4); 114-1150).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service