Z743432
Laser Cryo-Tags® sidewall
blue, size 1.50 in. × 0.25 in., pkg of 3120 labels 20 sheets of 156 labels
Synonym(s):
cryogenic plate labels, freezer labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41103032
NACRES:
NB.35
Recommended Products
packaging
pkg of 3120 labels 20 sheets of 156 labels
manufacturer/tradename
Diversified Biotech SIDE-1100
technique(s)
cryopreservation: suitable
size
1.50 in. × 0.25 in.
color
blue
Looking for similar products? Visit Product Comparison Guide
General description
Sidewall Cryo-Tags for laser printers withstand both long term cryogenic storage conditions and conventional freezer storage. These labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic multiwell plates. Cryo-Tags also withstand flash/snap freezing in liquid nitrogen, boiling water baths at 100°C and dry heat up to 150°C.
Legal Information
D-230 is a registered trademark of Texaco
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service